1-Methyl-5-(trifluoromethyl)pyrazole-4-carboxylic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 1-Methyl-5-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid

M2753-200MG M2753-1G
TCIAM2753-200MGEA 64 EUR
TCIAM2753-200MG TCIAM2753-1G
1-Methyl-5-(trifluoromethyl)pyrazole-4-carboxylic acid ≥98.0% (by GC, titration analysis)
1-Methyl-5-(trifluoromethyl)pyrazole-4-carboxylic acid
Formula: C₆H₅F₃N₂O₂
MW: 194,11 g/mol
Número MDL: MFCD04110182
Núm. CAS: 119083-00-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15647ae50d6d1b9b8555796f6ee30575a7087180&vid=15647ae50d6d1b9b8555796f6ee30575a7087180&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 121.0 to 125.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR