4-methoxy-1-naphthoic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 4-Methoxynaphthalene-1-carboxylic acid, 4-Methoxy-1-naphthoic acid

M2763-1G M2763-5G
TCIAM2763-1GEA 54 EUR
TCIAM2763-1G TCIAM2763-5G
4-methoxy-1-naphthoic acid ≥98.0% (by GC, titration analysis)
4-methoxy-1-naphthoic acid
Formula: C₁₂H₁₀O₃
MW: 202,209 g/mol
Número MDL: MFCD02575372
Núm. CAS: 13041-62-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48d1e6b890d3232c3fe9a6aecef114ddc6ac2e58&vid=48d1e6b890d3232c3fe9a6aecef114ddc6ac2e58&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 243.0 to 247.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR