Methyl 4-Acetamido-2-methoxybenzoate ≥98.0% (por GC)

Proveedor: TCI
Atención

Sinónimos: 4-Acetamido-o-anisic acid methyl ester, 4-Acetamido-2-methoxybenzoic acid methyl ester, Methyl 4-Acetamido-o-anisate, Methyl 4-acetamido-2-methoxybenzoate

M2776-5G M2776-25G
TCIAM2776-5GEA 18 EUR
TCIAM2776-5G TCIAM2776-25G
Methyl 4-Acetamido-2-methoxybenzoate ≥98.0% (por GC)
Methyl 4-Acetamido-2-methoxybenzoate
Formula: C₁₁H₁₃NO₄
MW: 223,23 g/mol
Número MDL: MFCD00065258
Núm. CAS: 4093-29-2
EINECS: 223-839-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a6a6e9f8bc50e51ceb05acf5e8aa8ca8168eb99&vid=5a6a6e9f8bc50e51ceb05acf5e8aa8ca8168eb99&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 128.0 to 132.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR