Methyl (3-bromophenyl)acetate ≥98.0% (por GC)

Proveedor: TCI

Sinónimos: 3-Bromophenylacetic acid methyll ester, Methyl 3-Bromophenylacetate, Methyl 2-(3-bromophenyl)acetate

M2804-5G M2804-25G
TCIAM2804-5GEA 35 EUR
TCIAM2804-5G TCIAM2804-25G
Methyl (3-bromophenyl)acetate ≥98.0% (por GC)
Methyl (3-bromophenyl)acetate
Formula: C₉H₉BrO₂
MW: 229,0700 g/mol
Número MDL: MFCD06858764
Núm. CAS: 150529-73-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea08fedbb8fa8442d04b636fd9886fea59f6ca3f&vid=ea08fedbb8fa8442d04b636fd9886fea59f6ca3f&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR