4-Methoxy-2-nitrobenzoic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 2-Nitro-p-anisic acid, 2-Nitro-4-methoxybenzoic acid

M2828-1G M2828-5G
TCIAM2828-1GEA 33 EUR
TCIAM2828-1G TCIAM2828-5G
4-Methoxy-2-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
4-Methoxy-2-nitrobenzoic acid
Formula: C₈H₇NO₅
MW: 197,15 g/mol
Número MDL: MFCD04972109
Núm. CAS: 33844-21-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c630ea427f5eaa12e60d2e822a9422f865a227e&vid=1c630ea427f5eaa12e60d2e822a9422f865a227e&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 198.0 to 202.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR