1-(4-Methoxyphenyl)cyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
1-(4-Methoxyphenyl)-1-cyclopropanecarboxylic acid
M2837-25G
M2837-5G
TCIAM2837-25GEA
429
EUR
TCIAM2837-25G
TCIAM2837-5G
1-(4-Methoxyphenyl)cyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)
1-(4-Methoxyphenyl)cyclopropanecarboxylic acid
Formula:
C₁₁H₁₂O₃ MW: 192,2112 g/mol |
Número MDL:
MFCD00019223 Núm. CAS: 16728-01-1 EINECS: 240-788-5 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 128.0 to 132.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...