4-Methoxy-3-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: (4-Methoxy-3-(trifluoromethyl)phenyl)boronic acid, 4-Methoxy-3-trifluoromethylphenylboronic acid, 4-Methoxy-3-(trifluoromethyl)benzeneboronic acid

M2839-200MG M2839-1G
TCIAM2839-200MGEA 60 EUR
TCIAM2839-200MG TCIAM2839-1G
4-Methoxy-3-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
4-Methoxy-3-(trifluoromethyl)phenylboronic acid
Formula: C₈H₈BF₃O₃
MW: 219,95 g/mol
Número MDL: MFCD07363790
Núm. CAS: 149507-36-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97723b233f32298abfbe56a56f7c5854c57b335c&vid=97723b233f32298abfbe56a56f7c5854c57b335c&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white crystal to powder
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) 97.0 to 109.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR