Methyl 3-Amino-2-fluorobenzoate ≥96.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Methyl 3-amino-2-fluorobenzoate, 3-Amino-2-fluorobenzoic acid methyl ester
M2868-200MG
M2868-1G
TCIAM2868-200MGEA
51
EUR
TCIAM2868-200MG
TCIAM2868-1G
Methyl 3-Amino-2-fluorobenzoate ≥96.0% (by GC, titration analysis)
Methyl 3-Amino-2-fluorobenzoate
Formula:
C₈H₈FNO₂ MW: 169,16 g/mol |
Número MDL:
MFCD12923265 Núm. CAS: 1195768-18-3 |
Resultados test especificaciones
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 96.0 % |
Purity (Nonaqueous Titration) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...