Methyl-6-fluorochromane-2-carboxylate ≥98.0% (por GC)

Proveedor: TCI

Sinónimos: Methyl 6-Fluorochroman-2-carboxylate, 6-Fluoro-3,4-dihydro-2H-1-benzopyran-2-carboxylic acid methyll ester, Methyl 6-Fluoro-3,4-dihydro-2H-1-benzopyran-2-carboxylate, 6-Fluorochroman-2-carboxylic acid methyll ester

M2879-1G M2879-5G
TCIAM2879-1GEA 30 EUR
TCIAM2879-1G TCIAM2879-5G
Methyl-6-fluorochromane-2-carboxylate ≥98.0% (por GC)
Methyl-6-fluorochromane-2-carboxylate
Formula: C₁₁H₁₁FO₃
MW: 210.21 g/mol
Número MDL: MFCD09701969
Núm. CAS: 874649-82-8

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 46.0 to 50.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR