Methyl-N-boc-piperidine-3-carboxylate ≥98.0% (por GC)

Proveedor: TCI

Sinónimos: 1-Boc-3-piperidinecarboxylic acid methyll ester, 1-tert-Butyl 3-Methyl Piperidine-1,3-dicarboxylate, 1-(tert-Butoxycarbonyl)-3-piperidinecarboxylic acid methyll ester, Piperidine-1,3-dicarboxylic acid 1-tert-Butyl 3-Methyl ester, Methyl 1-(tert-Butoxycarbonyl)-3-piperidinecarboxylate, Methyl 1-Boc-3-piperidinecarboxylate, Methyl N-Boc-piperidine-3-carboxylate

M2918-5G M2918-25G
TCIAM2918-5GEA 16 EUR
TCIAM2918-5G TCIAM2918-25G
Methyl-N-boc-piperidine-3-carboxylate ≥98.0% (por GC)
Methyl-N-boc-piperidine-3-carboxylate
Formula: C₁₂H₂₁NO₄
MW: 243.2994 g/mol
Número MDL: MFCD06795926
Núm. CAS: 148763-41-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e25d822a644be8aff98e40be94a710611dc1b6ee&vid=e25d822a644be8aff98e40be94a710611dc1b6ee&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 47.0 to 51.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR