Mono-2-(methacryloyloxy)ethyl Phthalate ≥90.0% (by titrimetric analysis) estabilizado
Proveedor: TCI
|
Atención
|
Sinónimos:
Phthalic acid Mono-2-(methacryloyloxy)ethyl ester, 2-[[2-(Methacryloyloxy)ethoxy]carbonyl]benzoic acid, Mono-2-(methacryloyloxy)ethyl Hydrogen Phthalate
Estabilización:
Stabilised with 4-methoxyphenol (MEHQ)
M2940-25G
M2940-500G
TCIAM2940-25GEA
16
EUR
TCIAM2940-25G
TCIAM2940-500G
Mono-2-(methacryloyloxy)ethyl Phthalate ≥90.0% (by titrimetric analysis) estabilizado
Mono-2-(methacryloyloxy)ethyl Phthalate
Formula:
C₁₄H₁₄O₆ MW: 278,26 g/mol |
Número MDL:
MFCD00066298 Núm. CAS: 27697-00-3 EINECS: 248-616-0 |
Resultados test especificaciones
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 85.0 % |
Purity (Neutralization titration) | min. 90.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...