3-Methoxy-5-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: 3-Methoxy-5-(trifluoromethyl)benzeneboronic acid, 3-Methoxy-5-trifluoromethylphenylboronic acid

M2943-1G M2943-5G
TCIAM2943-1GEA 95 EUR
TCIAM2943-1G TCIAM2943-5G
3-Methoxy-5-(trifluoromethyl)phenylboronic acid (contains varying amounts of Anhydride)
3-Methoxy-5-(trifluoromethyl)phenylboronic acid
Formula: C₈H₈BF₃O₃
MW: 219,95 g/mol
Número MDL: MFCD08056362
Núm. CAS: 871332-97-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebc42279fb6bba8e340129ef4cbcc4a29bcb6989&vid=ebc42279fb6bba8e340129ef4cbcc4a29bcb6989&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 109.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR