Methylammonium Hexafluorophosphate ≥98.0% (by total nitrogen basis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Methylamine Hexafluorophosphate
M2989-1G
M2989-5G
TCIAM2989-1GEA
64
EUR
TCIAM2989-1G
TCIAM2989-5G
Methylammonium Hexafluorophosphate ≥98.0% (by total nitrogen basis)
Methylammonium Hexafluorophosphate
Formula:
CH₆F₆NP MW: 177,03 g/mol Pto. de fusión: 176°C |
Núm. CAS:
28302-50-3 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...