Methylamine Thiocyanate ≥97.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: Methylammonium Thiocyanate

M2991-1G M2991-5G
TCIAM2991-1GEA 46 EUR
TCIAM2991-1G TCIAM2991-5G
Methylamine Thiocyanate ≥97.0% (by titrimetric analysis)
Methylamine Thiocyanate
Formula: C₂H₆N₂S
MW: 90,14 g/mol
Núm. CAS: 61540-63-4

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR