1-(2-Methoxyethyl)-1-methylpyrrolidinium bis(fluorosulphonyl)imide ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

M2998-1G M2998-5G
TCIAM2998-1GEA 85 EUR
TCIAM2998-1G TCIAM2998-5G
1-(2-Methoxyethyl)-1-methylpyrrolidinium bis(fluorosulphonyl)imide ≥98.0% (by titrimetric analysis)
1-(2-Methoxyethyl)-1-methylpyrrolidinium bis(fluorosulphonyl)imide
Formula: C₈H₁₈F₂N₂O₅S₂
MW: 324,36 g/mol
Número MDL: MFCD30749293
Núm. CAS: 1235234-47-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0661c1753b681e2f35ef713c82609c049aaa563d&vid=0661c1753b681e2f35ef713c82609c049aaa563d&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (Ion exchange titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR