4-Methylumbelliferyl-β-D-glucopyranoside ≥98.0% (by HPLC)
Proveedor: TCI
Sinónimos:
4-Methylumbelliferyl beta-D-Glucopyranoside, 4-Methylumbelliferyl β-D-Glucopyranoside, 4-MU-β-D-Glc, 4-Methyl-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2H-chromen-2-one, MUGlucosid
M3022-1G
M3022-250MG
TCIAM3022-1GEA
77
EUR
TCIAM3022-1G
TCIAM3022-250MG
4-Methylumbelliferyl-β-D-glucopyranoside ≥98.0% (by HPLC)
4-Methylumbelliferyl-beta-D-glucopyranoside
Formula:
C₁₆H₁₈O₈ MW: 338,312 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00063694 Núm. CAS: 18997-57-4 EINECS: 242-736-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Specific rotation [a]20/D | -97.5 to -108.0 deg (C=0.3, water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...