4-Methylumbelliferyl beta-D-ribofuranoside ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: 7-(((2S,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-4-methyl-2H-chromen-2-one, 4-Methylumbelliferyl β-D-Ribofuranoside

M3027-25MG M3027-100MG
TCIAM3027-25MGEA 129 EUR
TCIAM3027-25MG TCIAM3027-100MG
4-Methylumbelliferyl beta-D-ribofuranoside ≥98.0% (by HPLC)
4-Methylumbelliferyl beta-D-ribofuranoside
Formula: C₁₅H₁₆O₇
MW: 308,29 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD06796941
Núm. CAS: 195385-93-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cf2d5bf407bae5f864d9f7d50119ca63fe2ed26&vid=3cf2d5bf407bae5f864d9f7d50119ca63fe2ed26&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D -147 to -152 deg (C=0.15, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR