4-(Methoxycarbonyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Proveedor: TCI

Sinónimos: 4-(Methoxycarbonyl)benzeneboronic acid pinacol ester, Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

M3057-1G M3057-5G
TCIAM3057-1GEA 25 EUR
TCIAM3057-1G TCIAM3057-5G
4-(Methoxycarbonyl)phenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
4-(Methoxycarbonyl)phenylboronic acid pinacol ester
Formula: C₁₄H₁₉BO₄
MW: 262,11 g/mol
Número MDL: MFCD02179438
Núm. CAS: 171364-80-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52ccd2c79fb715199aa92a9d29d39d0551949452&vid=52ccd2c79fb715199aa92a9d29d39d0551949452&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 80.0 to 84.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR