2-Methoxy-N-(4-methoxyphenyl)aniline ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
M3067-1G
M3067-5G
TCIAM3067-1GEA
142
EUR
TCIAM3067-1G
TCIAM3067-5G
2-Methoxy-N-(4-methoxyphenyl)aniline ≥98.0% (by GC, titration analysis)
2-Methoxy-N-(4-methoxyphenyl)aniline
Formula:
C₁₄H₁₅NO₂ |
Número MDL:
MFCD27956479 Núm. CAS: 58751-07-8 |
|
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 69.0 to 73.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...