2-Methyl-4-(trifluoromethyl)thiazole-5-carboxylic acid ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
M3078-1G
M3078-5G
TCIAM3078-1GEA
18
EUR
TCIAM3078-1G
TCIAM3078-5G
2-Methyl-4-(trifluoromethyl)thiazole-5-carboxylic acid ≥98.0% (by GC, titration analysis)
2-Methyl-4-(trifluoromethyl)thiazole-5-carboxylic acid
Formula:
C₆H₄F₃NO₂S MW: 211,1600 g/mol |
Número MDL:
MFCD00173295 Núm. CAS: 117724-63-7 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...