4-Methyl-1,2,3-thiadiazole-5-carboxylic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

M3081-1G M3081-5G
TCIAM3081-1GEA 31 EUR
TCIAM3081-1G TCIAM3081-5G
4-Methyl-1,2,3-thiadiazole-5-carboxylic acid ≥98.0% (by GC, titration analysis)
4-Methyl-1,2,3-thiadiazole-5-carboxylic acid
Formula: C₄H₄N₂O₂S
MW: 144,15 g/mol
Número MDL: MFCD00173830
Núm. CAS: 18212-21-0

Order Now

Resultados test especificaciones

Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR