Mollugin ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: 6-Hydroxy-2,2-dimethyl-2H-benzo[h]chromene-5-carboxylic acid methyl ester, Methyl 6-Hydroxy-2,2-dimethyl-2H-benzo[h]chromene-5-carboxylate

M3091-10MG M3091-50MG
TCIAM3091-10MGEA 65 EUR
TCIAM3091-10MG TCIAM3091-50MG
Mollugin ≥98.0% (by HPLC)
Mollugin
Formula: C₁₇H₁₆O₄
MW: 284,31 g/mol
Temperatura de almacenaje: Nevera
Número MDL: MFCD02752331
Núm. CAS: 55481-88-4

Order Now

Resultados test especificaciones

Appearance White to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 130.0 to 134.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR