Methenolone Enanthate ≥98.0% (by HPLC)
Proveedor: TCI
M3092-1G
M3092-200MG
TCIAM3092-1GEA
264
EUR
TCIAM3092-1G
TCIAM3092-200MG
Methenolone Enanthate ≥98.0% (by HPLC)
Methenolone Enanthate
Formula:
C₂₇H₄₂O₃ |
Número MDL:
MFCD00864190 Núm. CAS: 303-42-4 EINECS: 206-141-6 Índice Merck: 14,05967 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 66.0 to 70.0 deg-C |
Specific rotation [a]20/D | +38.0 to +42.0 deg (C=1, CHCL3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...