1-Methyl-3-(4-sulphobutyl)imidazolium Hydrogen sulphate ≥95.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

Sinónimos: 1-butylsulphonic-3-methylimidazolium hydrogensulphate

M3120-5G M3120-25G
TCIAM3120-5GEA 99 EUR
TCIAM3120-5G TCIAM3120-25G
1-Methyl-3-(4-sulphobutyl)imidazolium Hydrogen sulphate ≥95.0% (by titrimetric analysis)
1-Methyl-3-(4-sulphobutyl)imidazolium Hydrogen sulphate
Formula: C₈H₁₆N₂O₇S₂
MW: 316,34 g/mol
Número MDL: MFCD12761445
Núm. CAS: 827320-59-2
UN: 3265
ADR: 8,II

Order Now

Resultados test especificaciones

Appearance Colorless to Light yellow clear liquid
Purity (Neutralization titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR