Methylamine cyanate ≥98.0% (by total nitrogen basis)

Proveedor: TCI
Atención

Sinónimos: Methylammonium Cyanate

M3134-1G M3134-5G
TCIAM3134-1GEA 58 EUR
TCIAM3134-1G TCIAM3134-5G
Methylamine cyanate ≥98.0% (by total nitrogen basis)
Methylamine cyanate
Formula: C₂H₆N₂O
MW: 74,08 g/mol
Núm. CAS: 63405-91-4

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR