Moxidectin ≥98.0% (by total nitrogen basis)

Proveedor: TCI
Peligro

Sinónimos: Moxidectin, (2R,2a'E,2a1'S,4Z,4'E,5S,6S,6'R,8'E,11'R,15'S,17a'R,20'R,20a'R)-2a1',20'-Dihydroxy-4-(methoxyimino)-5,6',8',19'-tetramethyl-6-((E)-4-methylpent-2-en-2-yl)-2a1',3,4,5,6,6',7',10',11',14',15',17a',20',20a'-tetradecahydro-2'H,17'H-spiro[pyran-2,13'-[11,15]methano[1,5]dioxacyclooctadecino[9,8,7-cd]benzofuran]-17'-one

M3136-1G M3136-250MG
TCIAM3136-1GEA 175 EUR
TCIAM3136-1G TCIAM3136-250MG
Moxidectin ≥98.0% (by total nitrogen basis)
Moxidectin
Formula: C₃₇H₅₃NO₈
MW: 639,83 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD00866560
Núm. CAS: 113507-06-5
UN: 2811
ADR: 6.1,III
Índice Merck: 14,06290

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 130.0 to 134.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR