5-Methoxypyrazine-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

M3181-200MG M3181-1G
TCIAM3181-200MGEA 30 EUR
TCIAM3181-200MG TCIAM3181-1G
5-Methoxypyrazine-2-carboxylic acid ≥98.0% (by HPLC, titration analysis)
5-Methoxypyrazine-2-carboxylic acid
Formula: C6H6N2O3
MW: 154,13 g/mol
Número MDL: MFCD10697773
Núm. CAS: 40155-42-8

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point (Decomposition) 196.0 to 200.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR