1-(4-Morpholinophenyl)-1-phenylprop-2-yn-1-ol ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

M3184-1G M3184-5G
TCIAM3184-1GEA 58 EUR
TCIAM3184-1G TCIAM3184-5G
1-(4-Morpholinophenyl)-1-phenylprop-2-yn-1-ol ≥98.0% (by HPLC, titration analysis)
1-(4-Morpholinophenyl)-1-phenylprop-2-yn-1-ol
Formula: C₁₉H₁₉NO₂
MW: 293,37 g/mol
Núm. CAS: 194940-93-7

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 158.0 to 162.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR