2-(4-Methoxyphenyl)ethylamine Hydroiodide ≥98.0% (by HPLC)

Proveedor: TCI
Atención

M3240-1G M3240-5G
TCIAM3240-1GEA 23 EUR
TCIAM3240-1G TCIAM3240-5G
2-(4-Methoxyphenyl)ethylamine Hydroiodide ≥98.0% (by HPLC)
2-(4-Methoxyphenyl)ethylamine Hydroiodide
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07907d5238d7929b324abc305113e0a8f2e4a890&vid=4a43a612761aa12c661553f59961c8358d637b13&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR