3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
4,4,5,5-Tetramethyl-2-[3-(4-Methyl-1-piperazinylmethyl)phenyl]-1,3,2-dioxaborolane, 1-Methyl-4-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)piperazine, 3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid Pinacol ester, 1-Methyl-4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperazine
M3245-1G
M3245-5G
TCIAM3245-1GEA
56
EUR
TCIAM3245-1G
TCIAM3245-5G
3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester
Formula:
C₁₈H₂₉BN₂O₂ MW: 316,25 g/mol |
Número MDL:
MFCD09266181 Núm. CAS: 883738-27-0 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | gt. 44.0 and le 48.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...