Methyl-1-aminocyclopropanecarboxylate hydrochloride ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: 1-Aminocyclopropane-1-carboxylic acid Methyl Ester Hydrochloride, 1-Aminocyclopropanecarboxylic acid methyll ester hydrochloride, Methyl 1-aminocyclopropanecarboxylate hydrochloride, 1-Aminocyclopropanecarboxylic acid methyl ester hydrochloride, Methyl 1-aminocyclopropanecarboxylate HCl

M3258-1G M3258-5G
TCIAM3258-1GEA 46 EUR
TCIAM3258-1G TCIAM3258-5G
Methyl-1-aminocyclopropanecarboxylate hydrochloride ≥98.0% (by titrimetric analysis)
Methyl-1-aminocyclopropanecarboxylate hydrochloride
Formula: C₅H₁₀ClNO₂
MW: 151,5914 g/mol
Pto. de fusión: 186°C
Temperatura de almacenaje: Nevera
Número MDL: MFCD00078879
Núm. CAS: 72784-42-0

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR