Morfolina iodhidrato ≥98.0% (by total nitrogen basis)

Proveedor: TCI
Atención

M3286-25G M3286-5G
TCIAM3286-25GEA 149 EUR
TCIAM3286-25G TCIAM3286-5G
Morfolina iodhidrato ≥98.0% (by total nitrogen basis)
Morfolina iodhidrato
Formula: C₄H₁₀INO
MW: 215.03 g/mol
Núm. CAS: 58464-45-2

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR