Sodium-1-naphthalenesulphonate ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
1-Naphthalenesulphonic acid sodium salt, Naphthalene-1-sulphonic acid sodium salt
N0015-25G
N0015-500G
N0015-100G
TCIAN0015-25GEA
29
EUR
TCIAN0015-25G
TCIAN0015-500G
TCIAN0015-100G
Sodium-1-naphthalenesulphonate ≥98.0% (by HPLC, titration analysis)
Sodium-1-naphthalenesulphonate
Formula:
C₁₀H₈O₃S MW: 230,21 g/mol |
Número MDL:
MFCD00064964 Núm. CAS: 130-14-3 EINECS: 204-976-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Ion exchange titration) | min. 98.0 % (calcd.on anh.substance) |
Water | max. 5.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...