Triethanolamine phosphate ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
2,2',2''-Nitrilotriethanol Phosphate
N0103-25G
N0103-500G
TCIAN0103-25GEA
45
EUR
TCIAN0103-25G
TCIAN0103-500G
Triethanolamine phosphate ≥98.0% (by titrimetric analysis)
Triethanolamine phosphate
Formula:
C₆H₁₅NO₃ MW: 247,18 g/mol |
Número MDL:
MFCD00054303 Núm. CAS: 10017-56-8 EINECS: 233-010-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 106.0 to 110.0 deg-C |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...