5-Nitrobarbituric acid trihydrate ≥98.0% (by titrimetric analysis)
Proveedor: TCI
N0128-25G
N0128-5G
TCIAN0128-25GEA
161
EUR
TCIAN0128-25G
TCIAN0128-5G
5-Nitrobarbituric acid trihydrate ≥98.0% (by titrimetric analysis)
5-Nitrobarbituric acid trihydrate
Formula:
C₄H₃N₃O₅·3H₂O |
Número MDL:
MFCD00006667 Núm. CAS: 480-68-2 EINECS: 207-557-0 Índice Merck: 14,06585 |
|
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (Neutralization titration) | min. 98.0 % (calcd.on anh.substance) |
Solubility in hot Water | almost transparency |
Water | 21.0 to 26.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...