3-Nitrobenzenesulphonic acid hydrate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Peligro

N0138-25G N0138-5G
TCIAN0138-25GEA 346 EUR
TCIAN0138-25G TCIAN0138-5G
3-Nitrobenzenesulphonic acid hydrate ≥98.0% (by HPLC, titration analysis)
3-Nitrobenzenesulphonic acid hydrate
Formula: C₆H₇NO₆S
MW: 221,19 g/mol
Número MDL: MFCD00064383
Núm. CAS: 98-47-5
EINECS: 202-671-7
UN: 2305
ADR: 8,II

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water max. 10.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR