Nitrofurazone ≥98.0% (by HPLC and UV method)
Proveedor: TCI
|
Atención
|
Sinónimos:
5-Nitrofurfural Semicarbazone, 2-((5-Nitrofuran-2-yl)methylene)hydrazinecarboxamide, 5-Nitro-2-Furaldehyde Semicarbazone
N0200-25G
N0200-250G
TCIAN0200-25GEA
16
EUR
TCIAN0200-25G
TCIAN0200-250G
Nitrofurazone ≥98.0% (by HPLC and UV method)
Nitrofurazone
Formula:
C₆H₆N₄O₄ MW: 198,14 g/mol |
Número MDL:
MFCD00003225 Núm. CAS: 59-87-0 EINECS: 200-443-1 Índice Merck: 14,06600 |
Resultados test especificaciones
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Solubility in N,N-DMF | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...