4-Hydroxy-3-nitroso-1-naphthalenesulphonic acid hydrate ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: 2-Nitroso-1-naphthol-4-sulphonic acid

N0269-25G N0269-5G
TCIAN0269-25GEA 165 EUR
TCIAN0269-25G TCIAN0269-5G
4-Hydroxy-3-nitroso-1-naphthalenesulphonic acid hydrate ≥98.0% (by titrimetric analysis)
4-Hydroxy-3-nitroso-1-naphthalenesulphonic acid hydrate
Formula: C₁₀H₇NO₅S
MW: 253,23 g/mol
Número MDL: MFCD00003933
Núm. CAS: 3682-32-4
EINECS: 222-964-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd4288d64c9aa78b8f5577cd661c2e39676f98ae&vid=bd4288d64c9aa78b8f5577cd661c2e39676f98ae&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Orange to Brown powder to crystal
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Solubility in Methanol almost transparency
Water max. 10 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR