3-Nonenoic acid ≥98.0% (by titrimetric analysis)
Proveedor: TCI
N0312-5ML
N0312-1ML
TCIAN0312-5MLEA
129
EUR
TCIAN0312-5ML
TCIAN0312-1ML
3-Nonenoic acid ≥98.0% (by titrimetric analysis)
3-Nonenoic acid
Resultados test especificaciones
Appearance | Colorless to Light yellow to Red clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...