5-Norbornene-2,3-dicarboxylic anhydride ≥97.0% (by titrimetric analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic anhydride, Carbic anhydride, 3a,4,7,7a-Tetrahydro-4,7-methanoisobenzofuran-1,3-dione
N0331-25G
N0331-500G
TCIAN0331-25GEA
17
EUR
TCIAN0331-25G
TCIAN0331-500G
5-Norbornene-2,3-dicarboxylic anhydride ≥97.0% (by titrimetric analysis)
5-Norbornene-2,3-dicarboxylic anhydride
Formula:
C₉H₈O₃ MW: 164,16 g/mol |
Número MDL:
MFCD00069013 Núm. CAS: 826-62-0 EINECS: 212-557-9 Índice Merck: 14,01796 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Methanolysis Method) | min. 97.0 % |
Melting point | 163.0 to 166.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...