Cholesterol pelargonate ≥93.0% (por GC)

Proveedor: TCI

Sinónimos: Cholesterol Nonanoate, Nonanoic Acid Cholesterol Ester, Pelargonic Acid Cholesterol Ester

N0347-25G N0347-500G
TCIAN0347-25GEA 33 EUR
TCIAN0347-25G TCIAN0347-500G
Cholesterol pelargonate ≥93.0% (por GC)
Cholesterol pelargonate
Formula: C₃₆H₆₂O₂
Número MDL: MFCD00003643
Núm. CAS: 1182-66-7
EINECS: 214-658-3

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 93.0 %
Mesomorphic range 74.0 to 82.0deg-C
Specific rotation [a]20/D -27.0 to -31.0 deg (C=1, THF)

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR