Naphthalene-1,4,5,8-tetracarboxylic dianhydride ≥97.0% (by HPLC)

Proveedor: TCI
Atención

Sinónimos: NTCDA, Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride, 1,4,5,8-Naphthalenetetracarboxylic dianhydride

N0369-250G N0369-25G
TCIAN0369-250GEA 545 EUR
TCIAN0369-250G TCIAN0369-25G
Naphthalene-1,4,5,8-tetracarboxylic dianhydride ≥97.0% (by HPLC)
Naphthalene-1,4,5,8-tetracarboxylic dianhydride
Formula: C₁₄H₄O₆
MW: 268,18 g/mol
Número MDL: MFCD00006915
Núm. CAS: 81-30-1
EINECS: 201-342-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51fc932c9f4a2352083c0c130169a583875d6306&vid=51fc932c9f4a2352083c0c130169a583875d6306&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Gray to Brown powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization back titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR