DL-Norphenylephrine hydrochloride ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

N0389-25G N0389-5G
TCIAN0389-25GEA 159 EUR
TCIAN0389-25G TCIAN0389-5G
DL-Norphenylephrine hydrochloride ≥98.0% (by titrimetric analysis)
DL-Norphenylephrine hydrochloride
Formula: C₈H₁₁NO₂
Número MDL: MFCD00012879
Núm. CAS: 4779-94-6
EINECS: 225-323-6
Índice Merck: 14,06699

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR