N-(2-Carboxyethyl)iminodiacetic acid ≥98.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
N-(2-carboxyethyl)iminodiaceticacid, Nitrilodiacetic-propionic Acid
N0408-25G
N0408-5G
TCIAN0408-25GEA
130
EUR
TCIAN0408-25G
TCIAN0408-5G
N-(2-Carboxyethyl)iminodiacetic acid ≥98.0% (by titrimetric analysis)
N-(2-Carboxyethyl)iminodiacetic acid
Formula:
C₇H₁₁NO₆ MW: 205,17 g/mol |
Número MDL:
MFCD00059634 Núm. CAS: 6245-75-6 EINECS: 228-360-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...