6-Nitrofluorescein (isomer II) ≥98.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
3',6'-Dihydroxy-6-nitro-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 6-Nitro-3',6'-dihydroxyspiro[isobenzofuran-1(3H),9'-(9H)xanthen]-3-one
N0445-1G
N0445-5G
TCIAN0445-1GEA
153
EUR
TCIAN0445-1G
TCIAN0445-5G
6-Nitrofluorescein (isomer II) ≥98.0% (by titrimetric analysis)
6-Nitrofluorescein (isomer II)
Formula:
C₂₀H₁₁NO₇ MW: 377,31 g/mol |
Número MDL:
MFCD00059694 Núm. CAS: 27402-68-2 |
Resultados test especificaciones
Appearance | Orange to Red powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Sodium hydroxide solution | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...