1,6-Naphthyridine ≥97.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
Pyrido[4,3-b]pyridine, 1,6-Diazanaphthalene
N0462-1G
N0462-100MG
TCIAN0462-1GEA
225
EUR
TCIAN0462-1G
TCIAN0462-100MG
1,6-Naphthyridine ≥97.0% (by titrimetric analysis)
1,6-Naphthyridine
Formula:
C₈H₆N₂ MW: 130,15 g/mol |
Número MDL:
MFCD00059750 Núm. CAS: 253-72-5 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...