4-Hydroxy-3-nitrobiphenyl
Proveedor: TCI
|
Atención
|
Sinónimos:
2-Nitro-4-phenylphenol, 3-Nitro-[1,1'-biphenyl]-4-ol, 3-Nitrobiphenyl-4-ol
N0560-25G
N0560-250G
TCIAN0560-25GEA
66
EUR
TCIAN0560-25G
TCIAN0560-250G
4-Hydroxy-3-nitrobiphenyl
4-Hydroxy-3-nitrobiphenyl
Formula:
C₁₂H₉NO₃ MW: 215,21 g/mol |
Número MDL:
MFCD00014694 Núm. CAS: 885-82-5 EINECS: 212-946-3 |
Resultados test especificaciones
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 67.0 to 70.0 deg-C |
Solubility in hot Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...