4-Nitrophenylethylamine hydrochloride ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

N0648-25G N0648-5G
TCIAN0648-25GEA 92 EUR
TCIAN0648-25G TCIAN0648-5G
4-Nitrophenylethylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Nitrophenylethylamine hydrochloride
Formula: C₈H₁₁ClN₂O₂
Número MDL: MFCD00012900
Núm. CAS: 29968-78-3
EINECS: 249-980-3

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Melting point 211.0 to 217.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR