Nω-Nitro-L-arginine ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: L-NNA, H-Arg(NO2)-OH

N0660-25G N0660-5G
TCIAN0660-25GEA 52 EUR
TCIAN0660-25G TCIAN0660-5G
Nω-Nitro-L-arginine ≥98.0% (by HPLC, titration analysis)
Nomega-Nitro-L-arginine
Formula: C₆H₁₃N₅O₄
MW: 219.20 g/mol
Número MDL: MFCD00007033
Núm. CAS: 2149-70-4
EINECS: 218-418-9

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +23.0 to +31.0 deg (C=2,2mol/L HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR