3-(2-Naphthyl)-D-alanine ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

N0665-1G N0665-5G
TCIAN0665-1GEA 50 EUR
TCIAN0665-1G TCIAN0665-5G
3-(2-Naphthyl)-D-alanine ≥98.0% (by HPLC, titration analysis)
3-(2-Naphthyl)-D-alanine
Formula: C₁₃H₁₃NO₂
Número MDL: MFCD00038546
Núm. CAS: 76985-09-6

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +11.0 to +14.0 deg (C=1, 1mol/L HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR